Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300022088.mm.1 |
NM_001080818.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022088.mm.1 |
NM_001080818.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022088.mm.1 |
NM_001173553.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022088.mm.1 |
NM_001173553.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022088.mm.1 |
TC0300002804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300022094.mm.1 |
NM_001080818.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022094.mm.1 |
NM_001080818.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022094.mm.1 |
NM_001173553.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022094.mm.1 |
NM_001173553.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022094.mm.1 |
TC0300002804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300022095.mm.1 |
NM_001080818.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022095.mm.1 |
NM_001080818.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022095.mm.1 |
NM_001173553.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022095.mm.1 |
NM_001173553.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022095.mm.1 |
TC0300002804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300022092.mm.1 |
NM_001080818.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022092.mm.1 |
NM_001080818.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022092.mm.1 |
NM_001173553.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022092.mm.1 |
NM_001173553.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022092.mm.1 |
TC0300002804.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300022093.mm.1 |
NM_001080818.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022093.mm.1 |
NM_001080818.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022093.mm.1 |
NM_001173553.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022093.mm.1 |
NM_001173553.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022093.mm.1 |
TC0300002804.mm.1 |
MTA TC |