Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080818.2 | NP_001074287.1 | 2 - 3 | 692 | 143 | |
NM_001173553.1 | NP_001167024.1 | 2 - 3 | 692 | 143 | ||
GenBank mRNA | AK043956.1 | - | 2 - 3 | 676 | 143 | |
AK048250.1 | - | 2 - 3 | 597 | 143 | ||
AK052560.1 | - | 2 - 3 | 683 | 143 | ||
AK142294.1 | - | 2 - 3 | 692 | 143 | ||
BC072644.1 | - | 2 - 3 | 646 | 30356962 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |