Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200009882.mm.1 |
NM_001164493.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009882.mm.1 |
NM_001164493.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009882.mm.1 |
TC1200001373.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009883.mm.1 |
NM_001164493.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009883.mm.1 |
NM_001164493.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009883.mm.1 |
TC1200001373.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009884.mm.1 |
NM_001164493.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009884.mm.1 |
NM_001164493.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009884.mm.1 |
TC1200001373.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009881.mm.1 |
NM_001164493.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009881.mm.1 |
NM_001164493.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009881.mm.1 |
TC1200001373.mm.1 |
MTA TC |