Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164493.1 | NP_001157965.1 | 10 - 11 | 2663 | 99 | |
GenBank mRNA | AK122572.1 | - | 10 - 11 | 2316 | 99 | |
BC065168.1 | - | 10 - 11 | 2663 | 99 | ||
BC138283.1 | - | 9 - 10 | 2019 | 8861302 | 99 | |
BC145748.1 | - | 9 - 10 | 2019 | 40131051 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |