Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289524.1 | NP_001276453.1 | 6 - 7 | 843 | 81 | |
NM_001289525.1 | NP_001276454.1 | 4 - 5 | 590 | 81 | ||
NM_024194.6 | NP_077156.2 | 6 - 7 | 846 | 81 | ||
GenBank mRNA | AK011726.1 | - | 6 - 7 | 827 | 81 | |
AK017814.1 | - | 6 - 7 | 815 | 81 | ||
AK034978.1 | - | 826 | 81 | |||
AK152244.1 | - | 4 - 5 | 571 | 81 | ||
AK163288.1 | - | 6 - 7 | 814 | 81 | ||
AK170052.1 | - | 6 - 7 | 823 | 81 | ||
BC002279.1 | - | 2 - 3 | 235 | 3497400 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |