Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081358.2 | NP_001074827.1 | 19 - 20 | 3184 | 76 | |
NM_001291452.1 | NP_001278381.1 | 19 - 20 | 3184 | 76 | ||
NM_001291453.1 | NP_001278382.1 | 19 - 20 | 3184 | 76 | ||
GenBank mRNA | AK122498.1 | - | 19 - 20 | 2319 | 76 | |
BC138450.1 | - | 19 - 20 | 3091 | 8861470 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |