Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290707.1 | NP_001277636.1 | 21 - 22 | 2559 | 66 | |
NM_011866.2 | NP_035996.2 | 20 - 21 | 2513 | 66 | ||
GenBank mRNA | AF110507.1 | - | 20 - 21 | 2384 | 66 | |
AK039249.1 | - | 21 - 22 | 2559 | 66 | ||
AK147690.1 | - | 20 - 21 | 2431 | 66 | ||
AK162221.1 | - | 20 - 21 | 2168 | 66 | ||
AK162804.1 | - | 18 - 19 | 2054 | 66 | ||
AK166310.1 | - | 12 - 13 | 1525 | 66 | ||
AY360383.1 | - | 20 - 21 | 2513 | 66 | ||
BC113201.1 | - | 20 - 21 | 2379 | 40048238 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |