Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mctp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024703.1 | NP_001019874.1 | 12 - 13 | 1772 | 88 | |
GenBank mRNA | BC086658.1 | - | 12 - 13 | 1772 | 30546156 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |