Array Name |
Probe IDs |
Targets |
Target Type |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
853461 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
962893 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
743571 |
AK005743.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
743571 |
AK005837.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
743571 |
AK006940.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
743571 |
AK015808.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
743571 |
AK076986.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
743571 |
AK132753.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
743571 |
BC048488.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
743571 |
BC061155.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
743571 |
D16301.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
743571 |
NM_007593.5 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
484244 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
472424 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
371895 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
824130 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
58074 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
602821 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
929798 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
293630 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
339662 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
757228 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
134604 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
576894 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
831326 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
896476 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
183238 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
517385 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
858223 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
26481 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
112264 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
38974 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P166886 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P168433 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2098805 |
NM_001190374.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2098805 |
AK081635.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2098805 |
AK220376.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06261371 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06261392 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P168433 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06261413 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06261823 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01562187 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06261473 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P166886 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P213077 |
AK020361.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P295621 |
NM_001190374.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P295621 |
AK081635.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P168433 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P06261392 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P166886 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P01562187 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277480 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277377 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277655 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277337 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278246 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277254 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278239 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278254 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06314996 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277272 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277399 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277351 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06046507 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278334 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278171 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278217 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278289 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277342 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277387 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06539429 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277584 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278386 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278201 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278335 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278423 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278183 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06191197 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277369 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278226 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278202 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277307 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277262 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278194 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278224 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278251 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278213 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277353 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06314994 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277410 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278242 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278219 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277318 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278269 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278272 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277279 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277277 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06464227 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277349 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278259 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277333 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277490 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05980107 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277382 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278266 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277299 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278206 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277287 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278191 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278267 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277381 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278388 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278418 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278282 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278234 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278164 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278437 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277365 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277475 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277368 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278322 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06539462 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277305 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278291 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277286 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278448 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278215 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277291 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277264 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05920846 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278200 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06006240 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278321 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06666083 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278293 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06227615 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277470 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277313 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277308 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06328646 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06058762 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05888077 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277358 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277589 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277348 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06104261 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278249 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278413 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277390 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278271 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278445 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06024586 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277656 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278281 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277340 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277344 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278337 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277290 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277345 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277588 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278162 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278297 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278447 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278211 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06024588 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277481 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277357 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277591 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277330 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277406 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278422 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278180 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277258 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277393 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278433 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277417 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278287 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278184 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277380 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277491 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277478 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278186 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278175 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06610880 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278195 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278270 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278288 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06191207 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278263 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278169 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277331 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277328 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278174 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06073039 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278159 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278238 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278204 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277361 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06471687 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277391 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06422243 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278320 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06475617 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06603555 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277477 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277397 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278244 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277363 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278250 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06550620 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06539437 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05953703 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278398 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277580 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278431 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278182 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277396 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278396 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278177 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277360 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278189 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277471 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06227331 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277233 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278256 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278395 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278176 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278414 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278412 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277283 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278284 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277324 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06464224 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277422 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06581803 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277347 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06471529 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06159720 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278442 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277469 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277255 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278416 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277251 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277404 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278260 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06519671 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06475609 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06391544 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277409 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278221 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278209 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278207 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05986175 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06471533 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278427 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277326 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06386113 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06391545 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277355 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278283 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277366 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277269 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278425 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278179 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278315 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278232 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277303 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277583 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277592 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278440 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06373751 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278198 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278258 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277403 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277486 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278208 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278277 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278328 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278330 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06104259 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277292 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277411 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278216 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06316914 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06539451 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277473 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06158864 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277401 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278318 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277270 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278384 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278391 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278197 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278173 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278252 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06223385 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278187 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277483 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278247 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278443 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277386 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278225 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277310 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277296 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277294 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277334 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277415 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278274 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278276 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277271 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06603556 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06183007 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277585 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278262 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277489 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277314 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05986192 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277322 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278331 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277413 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277267 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P06400618 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278166 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277275 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278160 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277419 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278161 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25277282 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278317 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P25278275 |
- |
- |