Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172742.2 | NP_766330.2 | 12 - 13 | 1314 | 62 | |
GenBank mRNA | BC041672.1 | - | 12 - 13 | 1286 | 62 | |
BC055074.1 | - | 10 - 11 | 978 | 30052208 | 62 | |
BC058196.1 | - | 5 - 6 | 416 | 62 | ||
CF734341.1 | - | 7 - 8 | 693 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |