Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033439.3 | NP_001028611.2 | 1 - 2 | 497 | 78 | |
NM_001252132.1 | NP_001239061.1 | 1 - 2 | 497 | 78 | ||
GenBank mRNA | AK129264.1 | - | 1 - 2 | 486 | 78 | |
AK143909.1 | - | 1 - 2 | 496 | 78 | ||
BB612742.1 | - | 1 - 2 | 499 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |