Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcoln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134160.1 | NP_598921.1 | 6 - 7 | 827 | 146 | |
GenBank mRNA | AF475086.1 | - | 6 - 7 | 827 | 146 | |
AK030819.1 | - | 6 - 7 | 947 | 146 | ||
AK033008.1 | - | 6 - 7 | 941 | 146 | ||
AK035029.1 | - | 6 - 7 | 948 | 146 | ||
AY083531.1 | - | 6 - 7 | 825 | 146 | ||
BC106856.2 | - | 6 - 7 | 827 | 40047039 | 146 | |
BC106857.2 | - | 6 - 7 | 827 | 40047042 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |