Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172601.3 | NP_766189.1 | 6 - 7 | 624 | 131 | |
GenBank mRNA | AB232586.1 | - | 6 - 7 | 470 | 131 | |
AK005230.1 | - | 6 - 7 | 527 | 131 | ||
AK038393.1 | - | 6 - 7 | 559 | 131 | ||
AK044202.1 | - | 6 - 7 | 501 | 131 | ||
AK160249.1 | - | 6 - 7 | 775 | 131 | ||
AK160351.1 | - | 6 - 7 | 490 | 131 | ||
BC033312.1 | - | 6 - 7 | 521 | 4481516 | 131 | |
BC046334.1 | - | 6 - 7 | 566 | 6492838 | 131 | |
BC054719.1 | - | 6 - 7 | 557 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |