Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178654.4 | NP_848769.2 | 13 - 14 | 2279 | 134 | |
GenBank mRNA | AK028469.1 | - | 13 - 14 | 2264 | 134 | |
AK046248.1 | - | 13 - 14 | 2196 | 134 | ||
AK049713.1 | - | 13 - 14 | 2266 | 134 | ||
AK083425.1 | - | 8 - 9 | 1077 | 134 | ||
AK133298.1 | - | 13 - 14 | 2268 | 134 | ||
AK171092.1 | - | 13 - 14 | 2210 | 134 | ||
BC052073.1 | - | 14 - 15 | 2202 | 6400440 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |