Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053080.3 | NP_444310.3 | 6 - 7 | 733 | 74 | |
GenBank mRNA | AF152359.1 | - | 6 - 7 | 688 | 74 | |
AF246711.1 | - | 6 - 7 | 681 | 74 | ||
AF280404.1 | - | 6 - 7 | 685 | 74 | ||
AK043530.1 | - | 6 - 7 | 730 | 74 | ||
AK086764.1 | - | 6 - 7 | 746 | 74 | ||
AK147006.1 | - | 6 - 7 | 734 | 74 | ||
BC026667.1 | - | 5 - 6 | 480 | 74 | ||
BC058277.1 | - | 6 - 7 | 677 | 6515355 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |