Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008830.2 | NP_032856.2 | 7 - 8 | 893 | 80 | |
GenBank mRNA | AK050345.1 | - | 6 - 7 | 846 | 80 | |
BC150687.1 | - | 7 - 8 | 763 | 9087598 | 80 | |
J03398.1 | - | 7 - 8 | 893 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |