Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026483.2 | NP_080759.2 | 4 - 5 | 1138 | 136 | |
GenBank mRNA | AK013337.1 | - | 4 - 5 | 1126 | 136 | |
BC018177.1 | - | 4 - 5 | 1132 | 136 | ||
BC049270.1 | - | 4 - 5 | 1114 | 3988645 | 136 | |
CK390971.1 | - | 3 - 4 | 537 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |