Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008877.3 | NP_032903.3 | 12 - 13 | 1645 | 79 | |
GenBank mRNA | AK132254.1 | - | 12 - 13 | 1614 | 79 | |
BC014773.1 | - | 12 - 13 | 1646 | 4193957 | 79 | |
BC057186.1 | - | 12 - 13 | 1620 | 5097123 | 79 | |
J04766.1 | - | 12 - 13 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |