Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pinx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028228.3 | NP_082504.1 | 2 - 3 | 253 | 117 | |
GenBank mRNA | AF321817.1 | - | 2 - 3 | 201 | 117 | |
AF421879.1 | - | 2 - 3 | 136 | 117 | ||
AK012057.1 | - | 2 - 3 | 231 | 117 | ||
BC065781.1 | - | 2 - 3 | 152 | 117 | ||
BC115636.1 | - | 2 - 3 | 165 | 40102952 | 117 | |
BC115637.1 | - | 2 - 3 | 163 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |