Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028525.2 | NP_082801.2 | 1 - 2 | 120 | 79 | |
GenBank mRNA | AB009660.1 | - | 1 - 2 | 157 | 79 | |
AK006746.1 | - | 1 - 2 | 119 | 79 | ||
BC049714.1 | - | 1 - 2 | 120 | 6741678 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |