Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1983383 |
BC049714.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1983383 |
NM_028525.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1983383 |
AB009660.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1983383 |
AK006746.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
873115 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
731452 |
AK011929.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
731452 |
AK014077.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
731452 |
BC028490.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
731452 |
NM_026523.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
432074 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
723196 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
857587 |
AK045060.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
857587 |
AK081454.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
268984 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
37321 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
126496 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P127144 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1983383 |
BC049714.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1983383 |
NM_028525.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1983383 |
AB009660.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1983383 |
AK006746.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665642 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665649 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665676 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665677 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665670 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665673 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665634 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665674 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665668 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665675 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665652 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665685 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665666 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665683 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665646 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665636 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665672 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665662 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665659 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665655 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665657 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665680 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665653 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665664 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665638 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665650 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665639 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665661 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665682 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29665648 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P127144 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08139171 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P164148 |
BC049714.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P164148 |
NM_028525.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P164148 |
AB009660.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P164148 |
AK006746.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P127144 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11229749 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11297561 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11297559 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11229745 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11229748 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665652 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665717 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665735 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665646 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665674 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665675 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665706 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665746 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665731 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665670 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665723 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665650 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665638 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665666 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665628 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665725 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665710 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665728 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665639 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665633 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665721 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665642 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665719 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665659 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665655 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665703 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665631 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665709 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665683 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665648 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665649 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665662 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665701 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665715 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665672 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665692 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665679 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665685 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665673 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665739 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665736 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665664 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665636 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665726 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665711 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665708 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665677 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665729 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665668 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665697 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665698 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665748 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665694 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29665644 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665670 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11488294 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11477996 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665636 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665711 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11595101 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32974869 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665691 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32974862 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11207796 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665747 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32974875 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11343880 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665724 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665708 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32974860 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665681 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665634 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665653 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32974872 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11514800 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665723 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32974868 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665729 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11599041 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11768136 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11229745 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11334405 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665652 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665717 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32974861 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665686 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665694 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11765075 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665633 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32974859 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665631 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665666 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665706 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665648 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665680 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665736 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32974854 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32974870 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11229748 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665748 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32974856 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11765076 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665715 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11488287 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665719 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32974863 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32974853 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11514801 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665663 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665692 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32974871 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665659 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665728 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665683 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32974876 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665655 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665725 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665697 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11619184 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665698 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665673 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665735 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32974874 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32974858 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11334390 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665703 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665726 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665739 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11156154 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665642 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11334400 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11297559 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32974865 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665639 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665668 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665674 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665682 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32974866 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11423590 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665625 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11334401 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665741 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P11163344 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665721 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29665646 |
- |
- |