Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mipep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027436.3 | NP_081712.2 | 17 - 18 | 2088 | 72 | |
NR_040642.1 | 17 - 18 | 2088 | 72 | |||
GenBank mRNA | AK017491.1 | - | 17 - 18 | 2064 | 72 | |
AK038479.1 | - | 17 - 18 | 2082 | 72 | ||
AK146428.1 | - | 18 - 19 | 2079 | 72 | ||
BC049871.1 | - | 8 - 9 | 928 | 72 | ||
BC110358.1 | - | 17 - 18 | 2066 | 5322591 | 72 | |
BC144798.1 | - | 17 - 18 | 2069 | 72 | ||
BC145711.1 | - | 17 - 18 | 2069 | 40131014 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |