Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec24b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207209.2 | NP_997092.1 | 13 - 14 | 2487 | 103 | |
GenBank mRNA | AK150076.1 | - | 12 - 13 | 1564 | 103 | |
AK152339.1 | - | 11 - 12 | 1459 | 103 | ||
BC046556.1 | - | 13 - 14 | 2319 | 103 | ||
BC046776.1 | - | 13 - 14 | 2487 | 5706007 | 103 | |
BC057355.1 | - | 13 - 14 | 2348 | 6833698 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |