Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020506.1 | NP_065252.1 | 5 - 6 | 573 | 72 | |
GenBank mRNA | AF145021.1 | - | 5 - 6 | 573 | 72 | |
AK084907.1 | - | 6 - 7 | 758 | 72 | ||
AK129430.1 | - | 5 - 6 | 568 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |