Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144803.2 | NP_659052.1 | 2 - 3 | 189 | 138 | |
GenBank mRNA | AY574261.1 | - | 1 - 2 | 73 | 138 | |
BC011490.1 | - | 2 - 3 | 189 | 4193582 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |