Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009618.2 | NP_033748.2 | 9 - 10 | 839 | 68 | |
GenBank mRNA | AK016550.1 | - | 9 - 10 | 839 | 68 | |
AK161261.1 | - | 9 - 10 | 802 | 68 | ||
AK161266.1 | - | 9 - 10 | 828 | 68 | ||
U16242.1 | - | 9 - 10 | 828 | 68 | ||
U38806.1 | - | 9 - 10 | 844 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mus musculus Probe spans exons |