Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199470.2 | NP_955764.1 | 9 - 10 | 75 | ||
GenBank mRNA | BC057373.1 | - | 6817232 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |