Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400015802.mm.1 |
NM_001039198.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015802.mm.1 |
NM_001039198.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015802.mm.1 |
TC1400002079.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015803.mm.1 |
NM_001039198.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015803.mm.1 |
NM_001039198.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015803.mm.1 |
TC1400002079.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015804.mm.1 |
NM_001039198.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015804.mm.1 |
NM_001039198.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015804.mm.1 |
TC1400002079.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015800.mm.1 |
NM_001039198.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015800.mm.1 |
NM_001039198.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015800.mm.1 |
TC1400002079.mm.1 |
MTA TC |