Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194346.2 | NP_919327.2 | 4 - 5 | 643 | 85 | |
GenBank mRNA | AK156320.1 | - | 4 - 5 | 726 | 85 | |
AK157962.1 | - | 4 - 5 | 689 | 85 | ||
AY283085.1 | - | 4 - 5 | 549 | 85 | ||
BC057595.1 | - | 4 - 5 | 643 | 6811051 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |