Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164503.1 | NP_001157975.1 | 2 - 3 | 265 | 66 | |
GenBank mRNA | AK158300.1 | - | 2 - 3 | 186 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |