Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd11b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044751.1 | NP_001038216.1 | 1 - 2 | 189 | 90 | |
NM_008288.2 | NP_032314.2 | 2 - 3 | 215 | 90 | ||
GenBank mRNA | AI099306.1 | - | 1 - 2 | 192 | 90 | |
AI182353.1 | - | 2 - 3 | 215 | 90 | ||
AK149518.1 | - | 1 - 2 | 187 | 90 | ||
AK165212.1 | - | 1 - 2 | 187 | 90 | ||
BC132364.1 | - | 2 - 3 | 155 | 40130641 | 90 | |
BC138780.1 | - | 2 - 3 | 155 | 8861802 | 90 | |
BC145415.1 | - | 2 - 3 | 155 | 9053973 | 90 | |
DQ089001.1 | - | 1 - 2 | 91 | 90 | ||
S75207.1 | - | 1 - 2 | 136 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |