Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf3ip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153137.4 | NP_694777.3 | 11 - 12 | 1475 | 63 | |
GenBank mRNA | AK161211.1 | - | 8 - 9 | 1085 | 63 | |
AY383616.1 | - | 9 - 10 | 1059 | 63 | ||
BC036561.1 | - | 11 - 12 | 1283 | 63 | ||
BC137605.1 | - | 9 - 10 | 1086 | 8860624 | 63 | |
BC171933.1 | - | 9 - 10 | 1086 | 9054217 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |