Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199020.1 | NP_001185949.1 | 7 - 8 | 1320 | 62 | |
NM_021421.4 | NP_067396.3 | 6 - 7 | 1455 | 62 | ||
NR_037578.1 | 6 - 7 | 1414 | 62 | |||
NR_037579.1 | 5 - 6 | 1129 | 62 | |||
GenBank mRNA | AK077039.1 | - | 6 - 7 | 1402 | 62 | |
AK088834.1 | - | 5 - 6 | 1088 | 62 | ||
AK135709.1 | - | 7 - 8 | 1317 | 62 | ||
AK158731.1 | - | 6 - 7 | 1471 | 62 | ||
AK166619.1 | - | 7 - 8 | 1319 | 62 | ||
AK169653.1 | - | 5 - 6 | 1039 | 62 | ||
AK178186.1 | - | 2 - 3 | 310 | 62 | ||
AK209212.1 | - | 2 - 3 | 310 | 62 | ||
BC024907.1 | - | 6 - 7 | 1415 | 2812124 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |