Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207214.3 | NP_997097.1 | 5 - 6 | 1041 | 91 | |
GenBank mRNA | AK134562.1 | - | 5 - 6 | 553 | 91 | |
AK152307.1 | - | 5 - 6 | 770 | 91 | ||
AK157473.1 | - | 5 - 6 | 736 | 91 | ||
AK164067.1 | - | 5 - 6 | 763 | 91 | ||
AK167145.1 | - | 5 - 6 | 729 | 91 | ||
AK170148.1 | - | 5 - 6 | 734 | 91 | ||
AK171012.1 | - | 5 - 6 | 759 | 91 | ||
BC049967.1 | - | 5 - 6 | 698 | 3672313 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |