Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207214.3 | NP_997097.1 | 8 - 9 | 1233 | 119 | |
GenBank mRNA | AK134562.1 | - | 8 - 9 | 745 | 119 | |
AK152307.1 | - | 8 - 9 | 962 | 119 | ||
AK157473.1 | - | 8 - 9 | 928 | 119 | ||
AK164067.1 | - | 8 - 9 | 955 | 119 | ||
AK167145.1 | - | 8 - 9 | 921 | 119 | ||
AK170148.1 | - | 8 - 9 | 926 | 119 | ||
AK171012.1 | - | 8 - 9 | 951 | 119 | ||
BC049967.1 | - | 8 - 9 | 890 | 3672313 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |