Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243792.1 | NP_001230721.1 | 6 - 7 | 1094 | 82 | |
NM_011935.3 | NP_036065.1 | 5 - 6 | 1032 | 82 | ||
GenBank mRNA | AB093266.1 | - | 6 - 7 | 1113 | 82 | |
AB221580.1 | - | 5 - 6 | 866 | 82 | ||
AF117254.1 | - | 5 - 6 | 929 | 82 | ||
AK052731.1 | - | 6 - 7 | 1073 | 82 | ||
AK140676.1 | - | 3 - 4 | 381 | 82 | ||
BC082324.1 | - | 5 - 6 | 1000 | 6850748 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Mammalian Gene Collection Mus musculus Probe spans exons |