Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029990.1 | NP_001025161.1 | 11 - 12 | 1106 | 59 | |
GenBank mRNA | AK155096.1 | - | 11 - 12 | 1025 | 59 | |
AK156895.1 | - | 10 - 11 | 1336 | 59 | ||
AK172330.1 | - | 10 - 11 | 1336 | 59 | ||
BC138944.1 | - | 11 - 12 | 1059 | 8861966 | 59 | |
BC145472.1 | - | 10 - 11 | 988 | 59 | ||
BC145473.1 | - | 9 - 10 | 1071 | 59 | ||
BC145474.1 | - | 10 - 11 | 988 | 59 | ||
CO043212.1 | - | 3 - 4 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |