Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400005906.mm.1 |
NM_019402.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400005906.mm.1 |
NM_019402.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400005906.mm.1 |
TC1400002832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400005915.mm.1 |
NM_019402.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400005915.mm.1 |
NM_019402.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400005915.mm.1 |
TC1400002832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400005905.mm.1 |
NM_019402.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400005905.mm.1 |
NM_019402.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400005905.mm.1 |
TC1400002832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400005913.mm.1 |
NM_019402.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400005913.mm.1 |
NM_019402.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400005913.mm.1 |
TC1400002832.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400005907.mm.1 |
NM_019402.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400005907.mm.1 |
NM_019402.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400005907.mm.1 |
TC1400002832.mm.1 |
MTA TC |