Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253759.1 | NP_001240688.1 | 12 - 13 | 1891 | 68 | |
NM_172813.3 | NP_766401.1 | 12 - 13 | 1891 | 68 | ||
GenBank mRNA | AK051821.1 | - | 12 - 13 | 1839 | 68 | |
BC052062.1 | - | 9 - 10 | 1402 | 5703279 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |