Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011465.4 | NP_035595.2 | 31 - 32 | 4595 | 66 | |
GenBank mRNA | AF093576.1 | - | 31 - 32 | 4503 | 66 | |
AK161449.1 | - | 2 - 3 | 236 | 66 | ||
BC150747.1 | - | 31 - 32 | 4460 | 9087658 | 66 | |
U87455.1 | - | 31 - 32 | 4575 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |