Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011465.4 | NP_035595.2 | 34 - 35 | 5033 | 62 | |
GenBank mRNA | AF093576.1 | - | 34 - 35 | 4941 | 62 | |
AK161449.1 | - | 5 - 6 | 674 | 62 | ||
BC150747.1 | - | 34 - 35 | 4898 | 9087658 | 62 | |
M10276.1 | - | 2 - 3 | 170 | 62 | ||
U87455.1 | - | 34 - 35 | 5013 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |