Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011465.4 | NP_035595.2 | 44 - 45 | 6566 | 76 | |
GenBank mRNA | AF093576.1 | - | 44 - 45 | 6474 | 76 | |
AK086017.1 | - | 1 - 2 | 206 | 76 | ||
AK161449.1 | - | 14 - 15 | 3492 | 76 | ||
BC150747.1 | - | 44 - 45 | 9087658 | 76 | ||
U87455.1 | - | 44 - 45 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |