Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc28a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004184.3 | NP_001004184.2 | 5 - 6 | 561 | 94 | |
GenBank mRNA | BC061230.1 | - | 5 - 6 | 561 | 30379959 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |