Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019951.1 | NP_064335.1 | 3 - 4 | 410 | 87 | |
GenBank mRNA | AB025405.1 | - | 3 - 4 | 410 | 87 | |
AK007449.1 | - | 3 - 4 | 408 | 87 | ||
AK169560.1 | - | 3 - 4 | 423 | 87 | ||
BC010484.1 | - | 3 - 4 | 402 | 3483051 | 87 | |
BC057885.1 | - | 3 - 4 | 602 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |