Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009755.3 | NP_033885.2 | 16 - 17 | 2475 | 68 | |
NR_033241.1 | 16 - 17 | 3170 | 68 | |||
GenBank mRNA | AK004995.2 | - | 16 - 17 | 2109 | 68 | |
AK147534.1 | - | 14 - 15 | 4086 | 68 | ||
AK154239.1 | - | 16 - 17 | 2477 | 68 | ||
AK158218.1 | - | 16 - 17 | 2468 | 68 | ||
BC003925.1 | - | 6 - 7 | 1499 | 68 | ||
BC053923.1 | - | 16 - 17 | 3061 | 68 | ||
BC063079.1 | - | 12 - 13 | 1605 | 68 | ||
BC066062.1 | - | 16 - 17 | 2403 | 6849066 | 68 | |
L24755.1 | - | 16 - 17 | 2252 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |