Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110162.1 | NP_001103632.1 | 11 - 12 | 1790 | 128 | |
NM_175384.4 | NP_780593.3 | 11 - 12 | 1493 | 128 | ||
GenBank mRNA | AK031313.1 | - | 7 - 8 | 935 | 128 | |
AK160223.1 | - | 11 - 12 | 1553 | 128 | ||
AK166952.1 | - | 11 - 12 | 1560 | 128 | ||
BC037136.1 | - | 11 - 12 | 1590 | 128 | ||
BC067009.1 | - | 11 - 12 | 1521 | 128 | ||
BC075682.1 | - | 11 - 12 | 1493 | 128 | ||
BC116298.1 | - | 7 - 8 | 935 | 40095010 | 128 | |
BC116299.1 | - | 7 - 8 | 935 | 40095011 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |