Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133857.3 | NP_598618.1 | 1 - 2 | 162 | 143 | |
GenBank mRNA | AK036876.1 | - | 1 - 2 | 163 | 143 | |
AK045953.1 | - | 1 - 2 | 225 | 143 | ||
BC096383.1 | - | 1 - 2 | 162 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |