Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133857.3 | NP_598618.1 | 13 - 14 | 2845 | 128 | |
GenBank mRNA | AK036876.1 | - | 13 - 14 | 2846 | 128 | |
BC022221.1 | - | 3 - 4 | 987 | 128 | ||
BC132339.1 | - | 12 - 13 | 2578 | 40130616 | 128 | |
BC145707.1 | - | 12 - 13 | 2578 | 40131010 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |