Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133857.3 | NP_598618.1 | 8 - 9 | 1504 | 63 | |
GenBank mRNA | AK036876.1 | - | 8 - 9 | 1505 | 63 | |
AK040332.1 | - | 9 - 10 | 1400 | 63 | ||
AK140453.1 | - | 8 - 9 | 1548 | 63 | ||
BC096383.1 | - | 8 - 9 | 64 | |||
BC132339.1 | - | 7 - 8 | 1237 | 40130616 | 63 | |
BC145707.1 | - | 7 - 8 | 1237 | 40131010 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |