Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016805.2 | NP_058085.2 | 10 - 11 | 2050 | 72 | |
GenBank mRNA | AF073992.1 | - | 10 - 11 | 1987 | 72 | |
AK037517.1 | - | 10 - 11 | 1993 | 72 | ||
AK089058.1 | - | 10 - 11 | 2051 | 72 | ||
AK145499.1 | - | 10 - 11 | 2053 | 72 | ||
AK145682.1 | - | 10 - 11 | 2053 | 72 | ||
AK159084.1 | - | 10 - 11 | 2019 | 72 | ||
AK159614.1 | - | 10 - 11 | 2055 | 72 | ||
AK159956.1 | - | 10 - 11 | 1992 | 72 | ||
AK165844.1 | - | 10 - 11 | 2050 | 72 | ||
AK167200.1 | - | 10 - 11 | 2053 | 72 | ||
AK168657.1 | - | 10 - 11 | 1992 | 72 | ||
AK207780.1 | - | 1 - 2 | 83 | 72 | ||
BC018353.1 | - | 10 - 11 | 1895 | 3584692 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |